missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13902-1-AP-150UL
This item is not returnable.
View return policy
Description
CXCL16 acts as a scavenger receptor on macrophages, which specifically binds to OxLDL (oxidized low density lipoprotein), suggesting that it may be involved in pathophysiology such as atherogenesis. Induces a strong chemotactic response. Induces calcium mobilization. Binds to CXCR6/Bonzo.Specifications
CXCL16 | |
Polyclonal | |
Unconjugated | |
CXCL16 | |
C X C motif chemokine 16, CXCL16, CXCLG16, SCYB16, Small inducible cytokine B16, SR PSOX, SRPSOX, Transmembrane chemokine CXCL16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
58191 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H2A7 | |
CXCL16 | |
CXCL16 Fusion Protein Ag4872 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CXCL16 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction