missing translation for 'onlineSavingsMsg'
Learn More

CXCL2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen 500-P130BT-1MG

 View more versions of this product

Product Code. 17864593

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-2 (CXCL2). Anti-Murine MIP-2 (CXCL2)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine MIP-2 (CXCL2) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine MIP-2 (CXCL2) (500-P130) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine MIP-2 (CXCL2). Western Blot: To detect mMIP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mMIP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P130BT-1MG will be provided as 2 x 500 μg

Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CXCL2
Polyclonal
Biotin
Cxcl2
C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2a; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; CXCL2; Cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO β; GRO2; GRO2 oncogene; GROb; GRObeta; gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROβ; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; Mgsa-b; MIP 2 a; MIP 2 alpha; MIP 2 a; Mip2; MIP-2; MIP2 a; MIP2 a; MIP2A; MIP-2a; MIP2-alpha; SB-251353; Scyb; Scyb2; small inducible cytokine subfamily, member 2
Rabbit
Antigen affinity chromatography
RUO
20310
-20°C
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P10889
Cxcl2
E.coli-derived Recombinant Murine MIP-2 (CXCL2)
2 x 500 μg
Primary
Mouse
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers