missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D3 Rabbit anti-Human, Mouse, Rat, Clone: 4C5C8, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | Cyclin D3 |
---|---|
Clone | 4C5C8 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18321914
|
Bio-Techne
NBP3-16309-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18353645
|
Bio-Techne
NBP3-16309-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin D3 Monoclonal antibody specifically detects Cyclin D3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Cyclin D3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Wnt Signaling Pathway | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
896 | |
IgG | |
Affinity purified |
4C5C8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 202-292 of human Cyclin D3 (P30281). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title