missing translation for 'onlineSavingsMsg'
Learn More

Cyclin T1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

1935.00 SEK - 4555.00 SEK

Specifications

Antigen Cyclin T1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18313185
View Documents
Bio-Techne
NBP3-02978-20UL
20 μL
1935.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18323084
View Documents
Bio-Techne
NBP3-02978-100UL
100 μL
4555.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclin T1 Polyclonal specifically detects Cyclin T1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Cyclin T1
Polyclonal
Purified
RUO
Human
CCNT, CCNT1, CCNTcyclin-T, CDK9-associated C-type protein, cyclin C-related protein, cyclin T1, cyclin T1b, Cyclin-T, cyclin-T1, CYCT1, HIVE1, human immunodeficiency virus type 1 (HIV-1) expression (elevated) 1, Human immunodeficiency virus-1 expression, subunit of positive elongation transcription factor b
A synthetic peptide corresponding to a sequence within amino acids 220-320 of human Cyclin T1 (NP_001231.2).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
Rabbit
Cell Cycle and Replication
PBS with 50% glycerol, pH7.3.
904
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.