missing translation for 'onlineSavingsMsg'
Learn More
Cycloheximide from Microbial, MP Biomedicals Product Code.: 11308509

Cycloheximide from Microbial, MP Biomedicals

Product Code. 11308509 Shop All MP Biomedical Products
25 g
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy

Product Code. 11308509

Brand: MP Biomedicals 0210018325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • Cycloheximide is used in protein synthesis in apoptosis
  • It is also used in plant research to study disease resistance and as an ethylene stimulant, useful in studies involving fruit and leaf production

TRUSTED_SUSTAINABILITY

Specifications

CAS 66-81-9
Melting Point 103°C to 115°C
Color White
Molecular Formula C15H23NO4
Linear Formula C15H23NO4
MDL Number MFCD00082346
Quantity 25 g
Synonym cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
Molecular Weight (g/mol) 281.35
PubChem CID 6197
ChEBI CHEBI:27641
Formula Weight 281.4
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material CYCLOHEXIMIDE from Microbial
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cycloheximide from Microbial, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.