missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl ethyl ketone, 99%, Thermo Scientific Chemicals

2645.00 SEK - 10375.00 SEK

Chemical Identifiers

CAS 1123-86-0
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00060814
InChI Key AMHOPTNGSNYSBL-UHFFFAOYSA-N
Synonym ethyl cyclohexyl ketone, cyclohexylethylketone, 1-propanone, 1-cyclohexyl, cyclohexyl ethyl ketone, ketone, cyclohexyl ethyl, 1-cyclohexyl-1-propanone, 1-cyclohexyl-propan-1-one, propanoylcyclohexane, 1-methylacetylcyclohexan, acmc-1c9ub
PubChem CID 70748
IUPAC Name 1-cyclohexylpropan-1-one
SMILES CCC(=O)C1CCCCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11454337
SDS View Documents
Thermo Scientific Alfa Aesar
B20409.06
5 g
2645.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11464337
SDS View Documents
Thermo Scientific Alfa Aesar
B20409.14
25 g
10375.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

Solubility
Soluble in organic solvents.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 214-380-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexyl ethyl ketone, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.