missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals

Product Code. 11354835
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 953-91-3
Molecular Formula C13H18O3S
Molecular Weight (g/mol) 254.344
MDL Number MFCD00014291
InChI Key OHHPZPDQZMUTCA-UHFFFAOYSA-N
Synonym cyclohexyl p-toluenesulfonate, cyclohexyl tosylate, cyclohexyl toluenesulfonate, cyclohexylp-toluenesulfonate, cyclohexyl p-toluenesulphonate, benzenesulfonic acid, 4-methyl-, cyclohexyl ester, p-toluenesulfonic acid, cyclohexyl ester, cyclohexyl-p-toluene sulfonate, benzenesulfonic acid, cyclohexyl ester, acmc-20aj06
PubChem CID 13722
IUPAC Name cyclohexyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2CCCCC2

Specifications

Melting Point 45°C to 46°C
Quantity 5 g
Beilstein 2217391
Formula Weight 254.35
Percent Purity 97%
Chemical Name or Material Cyclohexyl p-toluenesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.