missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals

709.00 SEK - 2765.00 SEK

Chemical Identifiers

CAS 931-51-1
Molecular Formula C6H11ClMg
Molecular Weight (g/mol) 142.91
MDL Number MFCD00003816
InChI Key WMJMABVHDMRMJA-UHFFFAOYSA-M
Synonym cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
PubChem CID 13605
IUPAC Name magnesium;cyclohexane;chloride
SMILES Cl[Mg]C1CCCCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15478047
View Documents
Thermo Scientific Alfa Aesar
H51165.AE
100 mL
709.00 SEK
100mL
Please to purchase this item. Need a web account? Register with us today!
15488047
View Documents
Thermo Scientific Alfa Aesar
H51165.AP
500 mL
2765.00 SEK
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

931-51-1
142.91
WMJMABVHDMRMJA-UHFFFAOYSA-M
13605
Cl[Mg]C1CCCCC1
C6H11ClMg
MFCD00003816
cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
magnesium;cyclohexane;chloride

Specifications

931-51-1
MFCD00003816
Air and moisture sensitive
It reacts violently with water.
Cl[Mg]C1CCCCC1
142.91
142.91
Cyclohexylmagnesium chloride
C6H11ClMg
UN2924
cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
WMJMABVHDMRMJA-UHFFFAOYSA-M
magnesium;cyclohexane;chloride
13605
1M in MeTHF
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Cyclohexylmagnesium chloride, 1M in MeTHF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 2-Methyltetrahydrofuran, Cyclohexylmagnesium chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.