missing translation for 'onlineSavingsMsg'
Learn More

Cyproheptadine hydrochloride, MP Biomedicals™

Product Code. 11404632
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyproheptadine is a Ca2+ channel blocker; 5-HT2 serotonin receptor antagonist; antihistamine, antipruritic. It is a migraine prophylactic

TRUSTED_SUSTAINABILITY

Specifications

CAS 41354-29-4
Color White
Molecular Formula C42H50Cl2N2O3
MDL Number MFCD27967225,MFCD00012538,MFCD00012538,MFCD27967225,MFCD00242817
Quantity 250 mg
Synonym Periactin hydrochloride, 1-Methyl-4-(5-dibenzo(a, e)cycloheptatrienylidene) piperidine hydrochloride
InChI Key ZEAUHIZSRUAMQG-UHFFFAOYSA-N
SMILES [H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12
IUPAC Name dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
Molecular Weight (g/mol) 701.77
PubChem CID 74764338
ChEBI CHEBI:650917
Formula Weight 323.88 (anhydrous)
Physical Form Powder
Chemical Name or Material Cyproheptadine hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.