missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | Cystatin B |
---|---|
Concentration | 0.49 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cystatin A (also designated STF1, STFA, stefin A or cystatin AS) and cystatin B (also designated PME, CST6, STFB, CPI-B, stefin B and liver thiol proteinase inhibitor) are thiol protease inhibitors that form complexes with Papain and the cathepsins B, H and L. Cystatin A, a cytoplasmic protein, is one of the precursor proteins of the cornified cell envelope in keratinocytes and plays a role in epidermal development and maintece. Cystatin B protects against intracellular proteases leaking out of lysosomes and is primarily expressed in heart, liver and kidney.Specifications
Cystatin B | |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P04080 | |
1476 | |
Cystatin B Fusion Protein Ag1262 | |
Primary | |
-20°C | |
CSTB |
0.49 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CPI B, CST6, CSTB, Cystatin B, cystatin B (stefin B), EPM1, PME, Stefin B, STFB | |
CSTB | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title