Learn More
Hettich™ Cyto Rotor for Benchtop Centrifuge, 12-place
Especially designed for use in cytology, recommended for laboratories working exclusively in this area.
Brand: Hettich™ 1615
This item is not returnable.
View return policy
Description
- Cyto Rotor, 12-place
- Works without a special cyto centrifuge
- 12-place cyto rotors fit in the benchtop centrifuge models ROTOFIX 32 A and UNIVERSAL 320
- Can be fitted and removed very easily without the need for tools
- Exceptional range of accessories
- Enables performance of all of the common centrifugation procedures used to yield cytological preparations
Specifications
Cyto Rotor | |
12,000 rpm | |
69 mm | |
35° |
16,582 x g | |
103 mm | |
Hettich Benchtop Centrifuges | |
12 |