missing translation for 'onlineSavingsMsg'
Learn More
D-Biotin, ≥97.5%, Cell Culture Reagent Grade, MP Biomedicals™ Product Code.: 11455092

D-Biotin, ≥97.5%, Cell Culture Reagent Grade, MP Biomedicals™

Product Code. 11455092 Shop All MP Biomedical Products
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11455092

Brand: MP Biomedicals 0219463405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Color White
Density 1.267 g/cm3(Lit.)
Assay Percent Range ≥97.5%
Quantity 5 g
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
Formula Weight 244.3
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material D-Biotin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Biotin, ≥97.5%, Cell Culture Reagent Grade, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.