missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Brown-Yellow to Yellow |
Loss on Drying | 1.0 % max. (105°C, 2 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98+% |
Packaging | Plastic bottle |
Quantity | 500 g |
Specific Rotation Condition | + 72.50 (20.00°C c=2,H2O,5hrs.) |
Specific Rotation | +70.0° to +73.0° (20°C, 589nm) (c=2.5, H2O) (after 3 hrs) |
Formula Weight | 215.64 |
Percent Purity | 98.0 to 102.0% (on dry substance) (Total base) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'