missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Pantothenic acid calcium salt, Cell culture reagent grade, MP Biomedicals™
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Quantity | 500 g |
Formula Weight | 238.3 |
Grade | Cell Culture Reagent |
Density | 0.5g/cm3 |
Chemical Name or Material | D-Pantothenic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'