missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate, 99%

1573.89 SEK - 3041.28 SEK

Chemical Identifiers

CAS 6974-32-9
Molecular Formula C28H24O9
Molecular Weight (g/mol) 504.48
MDL Number MFCD00005357
InChI Key GCZABPLTDYVJMP-CBUXHAPBSA-N
Synonym 1-o-acetyl-2,3,5-tri-o-benzoyl-beta-d-ribofuranose, beta-d-ribofuranose 1-acetate 2,3,5-tribenzoate, 2r,3r,4r,5s-5-acetyloxy-3,4-bis benzoyloxy oxolan-2-yl methyl benzoate, 1-acetyl-2,3,5-tribenzoyl-beta-d-ribofuranose, 1-acetyl-2,3,5-tribenzoy-b-d-ribofuranose, 1-o-acetyl-2,3,5,tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5s-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl methyl benzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxymethyl tetrahydrofuran-3,4-diyl dibenzoate, .beta.-d-ribofuranose, 1-acetate 2,3,5-tribenzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxy methyl tetrahydrofuran-3,4-diyl dibenzoate
PubChem CID 81455
IUPAC Name [(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
SMILES CC(=O)OC1C(C(C(O1)COC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10761441
View Documents Promotion Details
Thermo Scientific Chemicals
170110100
10 g Glass bottle
1573.89 SEK
10g
Please to purchase this item. Need a web account? Register with us today!
10513864
View Documents Promotion Details
Thermo Scientific Chemicals
170110250
25 g Glass bottle
3041.28 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6974-32-9
504.48
GCZABPLTDYVJMP-CBUXHAPBSA-N
81455
CC(=O)OC1C(C(C(O1)COC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
C28H24O9
MFCD00005357
1-o-acetyl-2,3,5-tri-o-benzoyl-beta-d-ribofuranose, beta-d-ribofuranose 1-acetate 2,3,5-tribenzoate, 2r,3r,4r,5s-5-acetyloxy-3,4-bis benzoyloxy oxolan-2-yl methyl benzoate, 1-acetyl-2,3,5-tribenzoyl-beta-d-ribofuranose, 1-acetyl-2,3,5-tribenzoy-b-d-ribofuranose, 1-o-acetyl-2,3,5,tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5s-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl methyl benzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxymethyl tetrahydrofuran-3,4-diyl dibenzoate, .beta.-d-ribofuranose, 1-acetate 2,3,5-tribenzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxy methyl tetrahydrofuran-3,4-diyl dibenzoate
[(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate

Specifications

6974-32-9
100.0
Brown to Green or Yellow
98.5% min. (HPLC)
MFCD00005357
+ 24.40
Solubility in water: insoluble
CC(=O)OC1C(C(C(O1)COC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
504.48
504.48
Liquid
98.5
128.0°C to 130.0°C
Authentic
C28H24O9
+ 24.40 (20.00°C c=1,pyridine)
1-o-acetyl-2,3,5-tri-o-benzoyl-beta-d-ribofuranose, beta-d-ribofuranose 1-acetate 2,3,5-tribenzoate, 2r,3r,4r,5s-5-acetyloxy-3,4-bis benzoyloxy oxolan-2-yl methyl benzoate, 1-acetyl-2,3,5-tribenzoyl-beta-d-ribofuranose, 1-acetyl-2,3,5-tribenzoy-b-d-ribofuranose, 1-o-acetyl-2,3,5,tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5s-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl methyl benzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxymethyl tetrahydrofuran-3,4-diyl dibenzoate, .beta.-d-ribofuranose, 1-acetate 2,3,5-tribenzoate, 2s,3r,4r,5r-2-acetoxy-5-benzoyloxy methyl tetrahydrofuran-3,4-diyl dibenzoate
GCZABPLTDYVJMP-CBUXHAPBSA-N
[(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
81455
99%
β-D-Ribofuranose 1-acetate 2, 3, 5-tribenzoate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 230-220-4

Product Identifier
  • ß-D-Ribofuranose 1-acetate 2,3,5-tribenzoate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only