missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98%, MP Biomedicals™
D-Sorbitol, Anhydrous, Cell Culture Reagent, ≥98% is used for inducing osmotic stress.
773.80 SEK - 2639.40 SEK
Chemical Identifiers
CAS | 50-70-4 |
---|---|
Molecular Formula | C6H14O6 |
Molecular Weight (g/mol) | 182.17 |
InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
Synonym | d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, glucarine, diakarmon, multitol, sorbilande |
PubChem CID | 5780 |
ChEBI | CHEBI:17924 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11432031
|
MP Biomedicals
0219474201 |
1 kg |
773.80 SEK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442031
|
MP Biomedicals
0219474205 |
5 kg |
2639.40 SEK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels.
- May be used to induce osmotic stress.
Chemical Identifiers
50-70-4 | |
182.17 | |
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, glucarine, diakarmon, multitol, sorbilande | |
CHEBI:17924 |
C6H14O6 | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
5780 |
Specifications
50-70-4 | |
1.49 g/cm3 at 59°F | |
Very High | |
C6H14O6 | |
Water Solubility: 2,750,000 mg/L at 30°C | |
−4 to 0 ° (c=2, water) | |
5780 | |
9.9X10-9 mm Hg at 25°C | |
Cell Culture Grade | |
<20.0 μS/cm |
230°F | |
7.0 | |
542°F | |
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, glucarine, diakarmon, multitol, sorbilande | |
FBPFZTCFMRRESA-UHFFFAOYNA-N | |
182.17 | |
CHEBI:17924 | |
≥98% | |
Powder |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.