missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol hexaacetate, 97%, Thermo Scientific™

780.00 SEK - 2529.00 SEK

Chemical Identifiers

CAS 7208-47-1
Molecular Formula C18H26O12
Molecular Weight (g/mol) 434.394
MDL Number MFCD00008715
InChI Key NJVBTKVPPOFGAT-XMTFNYHQSA-N
Synonym d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate
PubChem CID 9802952
IUPAC Name [(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate
SMILES CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15437369
SDS View Documents
Thermo Scientific Alfa Aesar
J66121.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15447369
SDS View Documents
Thermo Scientific Alfa Aesar
J66121.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7208-47-1
434.394
NJVBTKVPPOFGAT-XMTFNYHQSA-N
9802952
CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
C18H26O12
MFCD00008715
d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate
[(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate

Specifications

7208-47-1
White
MFCD00008715
NJVBTKVPPOFGAT-XMTFNYHQSA-N
[(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate
9802952
97%
D-Sorbitol hexaacetate
100°C to 104°C
C18H26O12
d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate
CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
434.394
434.39
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 230-588-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.