Learn More
Memmert™ DAkkS Factory Calibration for UN75plus
DAkkS factory calibration for a freely selectable temperature for Memmert Ovens.
Brand:  Memmert™ UN75plusE39696
Additional Details : Weight : 0.01200kg
Product Code. 17570536
  • 24760.00 SEK / Each
Estimated Shipment: 30-08-2024
to see stock.
Description
Description
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
UN75plus Oven
DAkkS Factory Calibration