Learn More
Debio 0932, MedChemExpress
MedChemExpress Debio 0932 (CUDC-305) is an orally active HSP90 inhibitor, with IC50s of 100 and 103 nM for HSP90α and HSP90β, respectively.
Brand:  Medchem Express HY-13469
Encompass_Preferred
 View more versions of this product
Product Code. 16421086
  • POA / 200mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Debio 0932 (CUDC-305) is an orally active HSP90 inhibitor, with IC50s of 100 and 103 nM for HSP90α and HSP90β, respectively.
Specifications
Specifications
Debio 0932
Off-White
C22H30N6O2S
CUDC-305
NC1=NC=CC2=C1N=C(SC3=C(N(C)C)C=C(OCO4)C4=C3)N2CCNCC(C)(C)C
442.58
Research
Cancer-programmed cell death
1061318-81-7
Research
200 mg
DMSO : 75 mg/mL (169.46 mM; Need ultrasonic)
442.58
98.29%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Debio 0932, MedChemExpress