missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borer Chemie™ deconex 22 LIQ-x
Universal use for automated cleaning of laboratory glassware and utensils. Without phosphates, surfactants and chlorine. deconex 22 LIQ-x is an ecological solution with an excellent performance. Acidic rinse with a deconex product recommended.
Brand: Borer Chemie™ 519200.00-K10W
This item is not returnable.
View return policy
Description
- Concentrate for universal automated cleaning
- Ecological solution without phosphates and surfactants
- Without chlorine, EDTA or NTA
- High yield, typical concentration: 5-10 mL/L
- Broad cleaning spectrum
- High performance thanks to high dispersing capacity
- Can be rinsed off without leaving residues
- For consistent and flawless results
- Acid rinse with a deconex cleaner recommended
- Technical support by the manufacturer
Specifications
Liquid | |
Alkalis, complexing agents, sequestering agents | |
Canister | |
1 x 12 kg | |
9.8 L | |
12 kg |
∼12.2 (1% Solution) | |
Concentrate | |
5°C to 25°C | |
2.6 gal. | |
26.5 lb. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- deconex 22 LIQ-x
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
- MIXTURE LIST-Contains : dipotassium oxo silicate, potassium hydroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Borer Chemie™ deconex 22 LIQ-x > deconex 22 LIQ-x; Quantity: 1 x 12 kg
Spot an opportunity for improvement?Share a Content Correction