Learn More
PEPperCHIP™ Dengue Virus Type 1 Proteome Microarray
Dengue Virus Type 1 Proteome Microarray with 3396 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
Brand: PEPperCHIP™ SPC.003.001
Additional Details : Weight : 0.02200kg
Description
- Microarray Content: Genome polyprotein (strain Singapore/S275/1990, UniProt ID P33478) translated into overlapping peptides
- Peptide Length/Overlap: 15 aa/ 14 aa
- Number of Peptides: 3396 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
PEPperSlide™ PEGMA/PMMA | |
Antibody validation, epitope mapping, serum screening, biomarker discovery | |
Microscopic Glass | |
Individual barcode for each microarray | |
1 Slide | |
1 mm | |
Transparent |
75.4 x 25 mm | |
High-density Peptide Microarray | |
Non-sterile | |
1 array copy per microarray | |
Combinatorial peptide synthesis by amino acid laser printing | |
ISO 9001:2015 | |
RUO |