Learn More
Tocris Bioscience™ DFHBI 1T
Mimic of GFP fluorophore for imaging RNA in living cells
Brand:  Tocris Bioscience™ 5610/10
Encompass
Product Code. 15736976
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
DFHBI 1T is a mimic of green fluorescent protein (GFP) fluorophore for imaging RNA in living cells. DFHBI 1T fluorescence is activated by binding to Spinach2 or Broccoli aptamers. DFHBI 1T exhibits peak excitation maxima of 482 nm and peak fluorescence emission of 505 nm when bound to Spinach2, enabling imaging with GFP filter cubes. Exhibits higher specific fluorescence and lower background fluorescence when bound to Spinach2 compared with Spinach2-DFHBI.
Specifications
Specifications
(5Z)-5-[(3,5-Difluoro-4-hydroxyphenyl)methylene]-3,5-dihydro-2-methyl-3-(2,2,2-trifluoroethyl)-4H-imidazol-4-one
1539318-36-9
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.