Learn More
Di-2-pyridyl thionocarbonate, 97%, Thermo Scientific Chemicals
861.50 SEK - 2804.60 SEK
Chemical Identifiers
CAS | 96989-50-3 |
---|---|
Molecular Formula | C11H8N2O2S |
Molecular Weight (g/mol) | 232.26 |
MDL Number | MFCD00074870 |
InChI Key | IKYOVSVBLHGFMA-UHFFFAOYSA-N |
Synonym | di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate |
PubChem CID | 719784 |
IUPAC Name | dipyridin-2-yloxymethanethione |
SMILES | C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10340304
|
Thermo Scientific Acros
390880010 |
1 g | Glass bottle |
861.50 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10259723
|
Thermo Scientific Acros
390880050 |
5 g | Glass bottle |
2804.60 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96989-50-3 | |
232.26 | |
IKYOVSVBLHGFMA-UHFFFAOYSA-N | |
719784 | |
C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2 |
C11H8N2O2S | |
MFCD00074870 | |
di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate | |
dipyridin-2-yloxymethanethione |
Specifications
96989-50-3 | |
Yellow to Brown | |
97% | |
MFCD00074870 | |
IKYOVSVBLHGFMA-UHFFFAOYSA-N | |
dipyridin-2-yloxymethanethione | |
719784 | |
97% | |
Di-2-pyridyl thionocarbonate |
96°C to 99°C | |
Authentic | |
C11H8N2O2S | |
di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate | |
C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2 | |
232.26 | |
232.26 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
- Di-2-pyridyl thionocarbonate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only