missing translation for 'onlineSavingsMsg'
Learn More

Di-2-thienyl ketoxime, 97%, Thermo Scientific™

Product Code. 11433707
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10558-44-8
Molecular Formula C9H7NOS2
Molecular Weight (g/mol) 209.28
MDL Number MFCD00173896
InChI Key UXSSDPITEOFAMK-UHFFFAOYSA-N
Synonym di thiophen-2-yl methanone oxime, di 2-thienyl methanone oxime, n-bis thiophen-2-yl methylidene hydroxylamine, dithiophen-2-ylmethanone oxime, di-2-thienylketoxim, maybridge1_002250, di2-thienylmethanone oxime, bis 2-thienyl ketoxime, methanone,di-2-thienyl-, oxime
PubChem CID 2775140
SMILES ON=C(C1=CC=CS1)C1=CC=CS1

Specifications

Melting Point 130°C to 134°C
Quantity 1 g
IUPAC Name N-(dithiophen-2-ylmethylidene)hydroxylamine
Formula Weight 209.29
Percent Purity 97%
Chemical Name or Material Di-2-thienyl ketoxime
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.