missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Lauroyl Peroxide, Pure, Fisher Chemical™
358.00 SEK - 615.00 SEK
Chemical Identifiers
CAS | 105-74-8 |
---|---|
Molecular Formula | C24H46O4 |
Molecular Weight (g/mol) | 398.628 |
MDL Number | 8964 |
InChI Key | YIVJZNGAASQVEM-UHFFFAOYSA-N |
Synonym | lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle |
PubChem CID | 7773 |
ChEBI | CHEBI:82479 |
IUPAC Name | dodecanoyl dodecaneperoxoate |
SMILES | CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC |
Description
Chemical Identifiers
105-74-8 | |
398.628 | |
YIVJZNGAASQVEM-UHFFFAOYSA-N | |
7773 | |
dodecanoyl dodecaneperoxoate |
C24H46O4 | |
8964 | |
lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle | |
CHEBI:82479 | |
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC |
Specifications
105-74-8 | |
White | |
8964 | |
YIVJZNGAASQVEM-UHFFFAOYSA-N | |
dodecanoyl dodecaneperoxoate | |
7773 | |
398.63g/mol |
53°C | |
C24H46O4 | |
lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle | |
CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC | |
398.628 | |
CHEBI:82479 | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dilauroyl peroxide
Signal Word
- Danger
Hazard Category
- Organic peroxide Type C & D
Hazard Statement
- H242-Heating may cause a fire.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P220-Keep/Store away from clothing/combustible materials.
- P234-Keep only in original container.
- P403+P235-Store in a well-ventilated place. Keep cool.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title