missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-hexyl Phthalate, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Di-n-hexyl phthalate | 84-75-3 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 84-75-3 |
Packaging | 2mL amber prescored glass ampule |
Quantity | 1 mL |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
Molecular Formula | C20H30O4 |
MDL Number | MFCD00043720 |
InChI Key | KCXZNSGUUQJJTR-UHFFFAOYSA-N |
SMILES | CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
IUPAC Name | 1,2-dihexyl benzene-1,2-dicarboxylate |
Show More |
Safety and Handling



Product Identifier
- DI-N-HEXYL PHTHALATE
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H360-May damage fertility or the unborn child.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : di-n-hexyl phthalate, methanol
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Di-n-hexyl Phthalate, SPEX CertiPrep™ >
Spot an opportunity for improvement?Share a Content Correction