missing translation for 'onlineSavingsMsg'
Learn More

Diallyl trisulfide, 98.8%, MP Biomedicals™

Product Code. 11422331
Click to view available options
Quantity:
25 mg
0.1 g
0.5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2050-87-5
Molecular Formula C6H10S3
Molecular Weight (g/mol) 178.326
InChI Key UBAXRAHSPKWNCX-UHFFFAOYSA-N
Synonym diallyl trisulfide, allyl trisulfide, allitridin, trisulfide, di-2-propenyl, diallyltrisulfide, diallyl trisulphide, allitridum, dasuansu, diallyltrisulfane, di-2-propenyl trisulfide
PubChem CID 16315
ChEBI CHEBI:78492
IUPAC Name 3-(prop-2-enyltrisulfanyl)prop-1-ene
SMILES C=CCSSSCC=C

Specifications

Color Colorless
Assay Percent Range 0.988
Quantity 0.1 g
Formula Weight 178.3
Physical Form Liquid
Chemical Name or Material DATS
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'