missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II), 97%, Thermo Scientific™

Product Code. 10763712
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1092372-91-2
Molecular Formula C20H48Cl2N2P2Ru
Molecular Weight (g/mol) 550.54
MDL Number MFCD07782003
InChI Key FUALRLYVUZSWKT-UHFFFAOYSA-L
Synonym dichlorobis 2-di-tert-butylphosphino ethylamine ruthenium ii, dichlorobis 2-di-t-butylphosphino ethylamine ruthenium ii
PubChem CID 71310624
IUPAC Name 2-ditert-butylphosphanylethanamine;dichlororuthenium
SMILES Cl[Ru]Cl.CC(C)(C)P(CCN)C(C)(C)C.CC(C)(C)P(CCN)C(C)(C)C

Specifications

Melting Point 247°C to 263°C
Color Brown to Yellow
Elemental Analysis 41.9 to 45.4% C, 4.9 to 5.3% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 550.54
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II)
Show More Show Less
compliance-icons
Product Identifier
  • Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II), 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.