missing translation for 'onlineSavingsMsg'
Learn More

Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals

Product Code. 15149263
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10210-68-1
Molecular Formula C8Co2O8
Molecular Weight (g/mol) 341.95
MDL Number MFCD00016024
Synonym carbon monoxide; cobalt 2+, dicobalt octacarbonyl stabilised with hexane, cobalt carbonyl, moistened with hexane hexane co

Specifications

Melting Point 51.0°C to 52.0°C
Color Orange-Red to Red
Density 1.8100g/mL
Flash Point −13°C
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass Bottle
Linear Formula Co2(CO)8
Quantity 100 g
UN Number 3124
Merck Index 15,3095
Specific Gravity 1.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and carbon disulfide
Stabilizer 5% max. hexane
Formula Weight 341.95
Percent Purity 95%
Physical Form Crystals or Flakes
Chemical Name or Material Dicobalt Octacarbonyl, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dicobalt octacarbonyl, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
  • Self-heating substance Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H251-Self-heating: may catch fire.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'