missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (2-aminoethyl)phosphonate oxalate, 97%, Thermo Scientific Chemicals

1505.50 SEK - 5508.76 SEK

Chemical Identifiers

CAS 203192-98-7
Molecular Formula C6H17NO3P
Molecular Weight (g/mol) 182.18
MDL Number MFCD00800256
InChI Key JUAGHBASZWRMQH-UHFFFAOYSA-O
Synonym diethyl 2-aminoethyl phosphonate oxalate, diethyl aminoethyl phosphonate oxalate, 2-diethoxyphosphoryl ethylamine oxalate, 2-diethoxyphosphorylethanamine; oxalic acid, oxalic acid diethyl 2-aminoethylphosphonate, diethyl 2-aminoethyl phosphonate oxalate salt, diethyl 2-aminoethyl phosphonate ethanedioate, aminoethylphosphonic acid diethylester oxalate salt
PubChem CID 2733595
IUPAC Name 2-diethoxyphosphorylethanamine;oxalic acid
SMILES CCOP(=O)(CC[NH3+])OCC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10621952
SDS View Documents
Thermo Scientific Acros
332220010
1 g Glass bottle
1505.50 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10135683
SDS View Documents
Thermo Scientific Acros
332220050
5 g Glass bottle
5508.76 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl (2-aminoethyl)phosphonate oxalate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.