missing translation for 'onlineSavingsMsg'
Learn More

Diethyl succinate, 98%, Thermo Scientific Chemicals

Product Code. 11462438
Click to view available options
Quantity:
10,000 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as emollients, masking agents, plasticisers and solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as emollients, masking agents, plasticisers and solvents.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-25-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00009208
InChI Key DKMROQRQHGEIOW-UHFFFAOYSA-N
Synonym diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377
PubChem CID 31249
IUPAC Name diethyl butanedioate
SMILES CCOC(=O)CCC(=O)OCC

Specifications

Melting Point -21°C
Density 1.043
Boiling Point 216°C to 218°C
Flash Point 90°C (194°F)
Refractive Index 1.42
Quantity 10,000 g
Beilstein 907645
Merck Index 14,8869
Solubility Information Slightly soluble in water.
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material Diethyl succinate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'