missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Diethyl Ether, ≥98%, Reagent Grade, Honeywell™

Product Code. 15608490
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Glass bottle
Poly bottle
Stainless steel can
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-36-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00009254
InChI Key RRQYJINTUHWNHW-UHFFFAOYSA-N
Synonym diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
PubChem CID 8179
ChEBI CHEBI:44664
IUPAC Name 1-ethoxy-2-(2-ethoxyethoxy)ethane
SMILES CCOCCOCCOCC

Specifications

Melting Point -44°C
Color Colorless
Density 0.909g/mL at 25°C
Boiling Point 189°C (1013 hPa)
Flash Point 67°C (152.6°F)
Odor Weak
Packaging Poly bottle
Linear Formula (C2H5OCH2CH2)2O
Quantity 4 L
Beilstein 1699259
Solubility Information Soluble in water at 20°C
Vapor Density 5.6 (vs air)
Formula Weight 162.23g/mol
Percent Purity ≥98%
Chemical Name or Material Diethylene glycol diethyl ether
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'