Learn More
Diethylphosphoramidic dichloride, 96%, Thermo Scientific™
Chemical Identifiers
CAS | 1498-54-0 |
---|---|
Molecular Formula | C4H10Cl2NOP |
Molecular Weight (g/mol) | 190.00 |
MDL Number | MFCD00013897 |
InChI Key | FXKRFCBTXGJQPY-UHFFFAOYSA-N |
Synonym | diethylphosphoramidic dichloride, n,n-diethylphosphoramidic dichloride, diethylphosphoramidoyl dichloride, phosphoramidic dichloride, diethyl, n,n-diethyl phosphoramidic dichloride, diethylaminophosphonoyl dichloride, n,n-dichloro-p,p-diethylphosphinic amide, n,p-diethylphosphinic amide, phosphoramidic dichloride, diethyl-8ci 9ci, diethylaminodichlorophosphine oxide |
PubChem CID | 73906 |
IUPAC Name | N-dichlorophosphoryl-N-ethylethanamine |
SMILES | CCN(CC)P(Cl)(Cl)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1498-54-0 | |
190.00 | |
FXKRFCBTXGJQPY-UHFFFAOYSA-N | |
73906 | |
CCN(CC)P(Cl)(Cl)=O |
C4H10Cl2NOP | |
MFCD00013897 | |
diethylphosphoramidic dichloride, n,n-diethylphosphoramidic dichloride, diethylphosphoramidoyl dichloride, phosphoramidic dichloride, diethyl, n,n-diethyl phosphoramidic dichloride, diethylaminophosphonoyl dichloride, n,n-dichloro-p,p-diethylphosphinic amide, n,p-diethylphosphinic amide, phosphoramidic dichloride, diethyl-8ci 9ci, diethylaminodichlorophosphine oxide | |
N-dichlorophosphoryl-N-ethylethanamine |
Specifications
1498-54-0 | |
Acidic | |
1.464 | |
UN3265 | |
diethylphosphoramidic dichloride, n,n-diethylphosphoramidic dichloride, diethylphosphoramidoyl dichloride, phosphoramidic dichloride, diethyl, n,n-diethyl phosphoramidic dichloride, diethylaminophosphonoyl dichloride, n,n-dichloro-p,p-diethylphosphinic amide, n,p-diethylphosphinic amide, phosphoramidic dichloride, diethyl-8ci 9ci, diethylaminodichlorophosphine oxide | |
CCN(CC)P(Cl)(Cl)=O | |
190.00 | |
190.01 | |
Diethylphosphoramidic dichloride |
220°C | |
C4H10Cl2NOP | |
MFCD00013897 | |
Moisture sensitive | |
FXKRFCBTXGJQPY-UHFFFAOYSA-N | |
N-dichlorophosphoryl-N-ethylethanamine | |
73906 | |
96% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.