missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Ketone, Extra Pure, Scharlab

Product Code. 11329953
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in analytical chemistry and in the synthesis of organic products.

  • Extra pure
  • Synonyms: 2,6-Dimethyl-4-heptanone, Isobutyl ketone
  • Molecular Formula: C9H180
  • Molar Mass: 142.24g/mol
  • CAS: 108-83-8
  • EIN ECS No.: 203-620-1
  • Solubility in water (20°C): 0.5g/l
  • Ignition temperature: 345°C
  • Vapor pressure: (20°C) 2.6hPa
  • Total isomer content (G.C.): Min 98%
  • Free acid (as CH3COOH): Max 0.01%
  • Copper (Cu): Max. 0.00002%
  • Iron (Fe): Max. 0.00005%
  • Lead (Pb): Max. 0.00002%
  • Nickel (Ni): Max. 0.00002%
  • Residue on Evaporation: Max. 0.005%

Specifications

CAS 108-83-8
Melting Point -46°C
Density 0.81g/cm3
Boiling Point 168°C
Flash Point 49°C
Molecular Formula C9H18O
Refractive Index 1.4143 at 20°C
Quantity 1 L
Synonym 2,6-dimethyl-4-heptanone, diisobutyl ketone, isovalerone, isobutyl ketone, valerone, diisobutylketone, diisobutilchetone, s-diisopropylacetone, di-isobutylcetone, dibk
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
SMILES CC(C)CC(=O)CC(C)C
IUPAC Name 2,6-dimethylheptan-4-one
Molecular Weight (g/mol) 142.242
PubChem CID 7958
Formula Weight 142.24g/mol
Grade Extra Pure
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisobutyl ketone, extra pure
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'