missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisodecyl Phthalate, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Specifications
Specifications
CAS | 26761-40-0 |
Boiling Range | 64°C |
Density | 0.79g/cm3 |
Boiling Point | 64°C |
Molecular Formula | C28H46O4 |
Quantity | 1 mL |
Synonym | diisodecyl phthalate, bis 8-methylnonyl phthalate, plasticized ddp, palatinol z, vestinol dz, didp plasticizer, bis isodecyl phthalate, di i-decyl phthalate, di-iso-decyl phthalate, sicol 184 |
InChI Key | ZVFDTKUVRCTHQE-UHFFFAOYSA-N |
Molecular Weight (g/mol) | 446.67 |
PubChem CID | 33599 |
Show More |
Safety and Handling
Product Identifier
- Diisodecyl phthalate
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction