Learn More
Dimethyl (2-oxopropyl)phosphonate, 95%, Thermo Scientific Chemicals
3033.65 SEK valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless to Brown |
Boiling Point | 76°C to 79°C (3.0 mmHg) |
Flash Point | 93°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Linear Formula | CH3COCH2P(O)(OCH3)2 |
Refractive Index | 1.4380 to 1.441 |
Quantity | 25 g |
Formula Weight | 166.11 |