missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylacetonitrile, MP Biomedicals™
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 86-29-3 |
Molecular Formula | C14H11N |
Quantity | 100 g |
Synonym | diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril |
InChI Key | NEBPTMCRLHKPOB-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2 |
IUPAC Name | 2,2-diphenylacetonitrile |
Molecular Weight (g/mol) | 193.249 |
PubChem CID | 6837 |
Formula Weight | 193.2 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'