missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphinic chloride, 97+%, Thermo Scientific Chemicals

Artikelnummer. 11409243
Click to view available options
Quantity:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.

Solubility
Decomposes with water.

Notes
Moisture Sensitive. Store away from strong bases, moisture. Incompatible with oxidizing agents,, Amines, Alcohols.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1499-21-4
Molecular Formula C12H10ClOP
Molecular Weight (g/mol) 236.63
MDL Number MFCD00002077
InChI Key QPQGTZMAQRXCJW-UHFFFAOYSA-N
Synonym diphenylphosphinic chloride, diphenylphosphinyl chloride, phosphinic chloride, diphenyl, diphenylphosphinoyl chloride, chlorodiphenylphosphine oxide, dpp-cl, unii-5gp30gy04v, diphenyl phosphinic chloride, diphenylphosphinochloridic acid, diphenylphosphinic acid chloride
PubChem CID 73910
IUPAC Name [chloro(phenyl)phosphoryl]benzene
SMILES ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Spezifikation

Melting Point ∼20°C
Density 1.242
Boiling Point 210°C to 212°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.609
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Decomposes with water.
Formula Weight 236.64
Percent Purity ≥97%
Chemical Name or Material Diphenylphosphinic chloride
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'