Learn More
Diquafosol tetrasodium, MedChemExpress
MedChemExpress Diquafosol tetrasodium is a P2Y2 receptor agonist that stimulates fluid and mucin secretion on the ocular surface, as a topical treatment of dry eye disease.
Brand:  Medchem Express HY-B0606
Encompass_Preferred
 View more versions of this product
Product Code. 16448386
  • 9647.57 SEK / 100mg
Explore more special offers
Description
Description
Diquafosol tetrasodium is a P2Y2 receptor agonist that stimulates fluid and mucin secretion on the ocular surface, as a topical treatment of dry eye disease.
Specifications
Specifications
Diquafosol tetrasodium
White
C18H22N4Na4O23P4
INS365
O[C@H]1[C@H](N(C=CC2=O)C(N2)=O)O[C@H](COP(OP(OP(OP(OC[C@H]3O[C@@H](N(C=CC4=O)C(N4)=O)[C@H](O)[C@@H]3O)(O[Na])=O)(O[Na])=O)(O[Na])=O)(O[Na])=O)[C@H]1O
878.23
Research
COVID-19-immunoregulation
211427-08-6
Research
100 mg
H2O : 100 mg/mL (113.87 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.14 mM; Need ultrasonic)
878.23
98.49%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.