Learn More
Diquafosol tetrasodium, MedChemExpress
MedChemExpress Diquafosol tetrasodium is a P2Y2 receptor agonist that stimulates fluid and mucin secretion on the ocular surface, as a topical treatment of dry eye disease.
Brand: Medchem Express HY-B0606
This item is not returnable.
View return policy
Description
Diquafosol tetrasodium is a P2Y2 receptor agonist that stimulates fluid and mucin secretion on the ocular surface, as a topical treatment of dry eye disease.Specifications
Diquafosol tetrasodium | |
White | |
C18H22N4Na4O23P4 | |
INS365 | |
O[C@H]1[C@H](N(C=CC2=O)C(N2)=O)O[C@H](COP(OP(OP(OP(OC[C@H]3O[C@@H](N(C=CC4=O)C(N4)=O)[C@H](O)[C@@H]3O)(O[Na])=O)(O[Na])=O)(O[Na])=O)(O[Na])=O)[C@H]1O | |
878.23 | |
Research | |
COVID-19-immunoregulation |
211427-08-6 | |
Research | |
100 mg | |
H2O : 100 mg/mL (113.87 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.14 mM; Need ultrasonic) | |
878.23 | |
98.49% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.