missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM MERC Biotech Disposable Sample Tubes for Osmometers
Disposable sample tube made from certified Medical Grade plastic, and consistently quality controlled on models 3900 and 3250 osmometers. For ease of access during use the box is fitted with a magnet opening mechanism.
Brand: GM MERC Biotech 101911
This item is not returnable.
View return policy
Description
- Certified Medical Grade plastic
- Manufactured in Denmark
- 0.2 mL to 0.3 mL sample size
- Designed for Osmometers Model 3900, Model 3250 and Model 3D3
- Consistently quality controlled on the 3900 and 3250 osmometers
Specifications
Sample Tube | |
Osmometers 3D3, 3250 and 3900 | |
Single-use | |
0.2 to 0.3 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction