Learn More
Kartell™ Disposable Stirrer for Cuvets
Manufactured from polystyrene to avoid breakage of fragile cuvets. Kartell™ Disposable Stirrers for Cuvets are designed with a rounded end for comfort and a flat stirring paddle for optimum mixing.
Brand: Kartell™ 0093900
Additional Details : Weight : 0.18000kg
Description
Manufactured from polystyrene to avoid breakage of fragile cuvets. Kartell™ Disposable Stirrers for Cuvets are designed with a rounded end for comfort and a flat stirring paddle for optimum mixing.
- CE-IVD certified
- Disposable
Specifications
3 mm | |
Polystyrene | |
90 mm |
Stirrer | |
CE-IVD |