missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dithranol, 99%, MP Biomedicals™
Click to view available options
Quantity:
0.25 g
1 g
5 g
Description
- Dithranol, a potential anti-psoriasis medication, may be used to study its safety, efficacy, metabolism and pharmacological profile.
- It may be used to study is physicochemical properties, as a reference compound and to develop effective drug delivery vehicles.
- It may be used as a matrix for matrix assisted laser desorption/ionization imaging on a Fourier transform ion cyclotron resonance mass spectrometer.
- Anthralin is a leukotriene biosynthesis inhibitor.
- Inhibits LTB4 ω-oxidation and disrupts mitochondria function.
- Anthralin is used in the treatment of psoriasis, as a fungicide, in the treatment of ringworm infections and in chronic dermatoses.
Specifications
Color | Orange-Yellow or Yellow |
Assay Percent Range | ≥99% |
Quantity | 5 g |
Synonym | anthralin, dithranol, 1,8-dihydroxyanthrone, 1,8-dihydroxy-9-anthrone, chrysodermol, cigthranol, batridol, cignolin, psoriacid-stift, 9 10h-anthracenone, 1,8-dihydroxy |
Formula Weight | 226.23 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Dithranol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'