Learn More
Elma™ Dividing Wall Sets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
Brand: Elma™ 1125222
This item is not returnable.
View return policy
Description
- For vertical division of the modular basket
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Specifications
Dividing Wall | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY | |
90 x 200 x 1 mm |
60 mm | |
Stainless Steel |