missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Octanol, 97%, Thermo Scientific Chemicals

Product Code. 10183510
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-96-6
MDL Number MFCD00004591
InChI Key SJWFXCIHNDVPSH-UHFFFAOYSA-N
Synonym 2-octanol, 2-octyl alcohol, hexylmethylcarbinol, 2-hydroxyoctane, sec-caprylic alcohol, 1-methylheptanol, methylhexylcarbinol, 1-methylheptyl alcohol, dl-2-octanol, n-octan-2-ol
PubChem CID 20083
ChEBI CHEBI:37869
IUPAC Name octan-2-ol
SMILES CCCCCCC(C)O

Specifications

Color Colorless to Yellow
Density 0.81g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H18O
Linear Formula CH3(CH2)5CH(OH)CH3
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Quantity 5 mL
Beilstein 01, 419
Fieser 01,109
Merck Index 15, 6841
Molecular Weight (g/mol) 130.231
Formula Weight 130.23
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material DL-2-Octanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'