missing translation for 'onlineSavingsMsg'
Learn More

DL-Epinephrine Crystalline MP Biomedicals

Product Code. 11406852
Click to view available options
Quantity:
1 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 329-65-7
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
InChI Key UCTWMZQNUQWSLP-UHFFFAOYSA-N
Synonym dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol
PubChem CID 838
ChEBI CHEBI:33568
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Quantity 100 g
Formula Weight 183.2
Physical Form Crystalline
Chemical Name or Material DL-Epinephrine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'