Learn More
Tocris Bioscience™ DMH2
Type I BMP receptor inhibitor
Brand: Tocris Bioscience™ 5580/50
This item is not returnable.
View return policy
Description
DMH2 is a type I BMP receptor inhibitor (Ki values are <1, 5.4 and 43 nM for ALK6, ALK3 and ALK2, respectively). Exhibits >30-fold selectivity over ALK4, ALK5, BMPR2, AMPK and VEGFR2. Also binds to TGFBR2 (Ki = 85 nM). Decreases growth and induces cell death of H1299 non-small cell lung carcinoma cells in vitro. Enhances liver regeneration in vivo.Specifications
4-[6-[4-[2-(4-Morpholinyl)ethoxy]phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline | |
1206711-14-9 | |
50 mg |