missing translation for 'onlineSavingsMsg'
Learn More

DNA Polymerase epsilon subunit 3 Mouse anti-Human, Clone: PCRP-POLE3-2F10, Novus Biologicals™

Mouse Monoclonal Antibody

2450.00 SEK

Specifications

Antigen DNA Polymerase epsilon subunit 3
Clone PCRP-POLE3-2F10
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18321642
View Documents
Bio-Techne
NBP3-07616-20UG
20 μg
2450.00 SEK
20µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNA Polymerase epsilon subunit 3 Monoclonal specifically detects DNA Polymerase epsilon subunit 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

DNA Polymerase epsilon subunit 3
0.2 mg/ml
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Chromatin Research, DNA Polymerases, DNA Repair
10 mM PBS with 0.05% BSA
54107
IgG1
Protein A or G purified
PCRP-POLE3-2F10
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array
Monoclonal
Purified
RUO
Human
Arsenic-transactivated protein, asTP, CHARAC17arsenic transactivated protein, CHRAC-17, CHRAC17Ybl1, Chromatin accessibility complex 17 kDa protein, DNA polymerase epsilon p17 subunit, DNA polymerase epsilon subunit 3, DNA polymerase epsilon subunit p17, DNA polymerase II subunit 3, EC 2.7.7.7, histone fold protein CHRAC17, HuCHRAC17, p17, polymerase (DNA directed), epsilon 3 (p17 subunit), YBL1
Recombinant full-length human DNA Polymerase epsilon subunit 3 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.