missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT1 Rabbit anti-Human, Mouse, Rat, Clone: 8E3H1, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | DNMT1 |
---|---|
Clone | 8E3H1 |
Dilution | Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:500 - 1:1000, Knockout Validated |
Applications | ChIP Assay, Immunofluorescence, Immunoprecipitation, KnockDown |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18322085
|
Bio-Techne
NBP3-15856-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18396963
|
Bio-Techne
NBP3-15856-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNMT1 Monoclonal antibody specifically detects DNMT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunofluorescence, Immunoprecipitation, KnockDownSpecifications
DNMT1 | |
Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:500 - 1:1000, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DNMT1 (NP_001370.1). MPARTAPARVPTLAVPAISLPDDVRRRLKDLERDSLTEKECVKEKLNLLHEFLQTEIKNQLCDLETKLRKEELSEEGYLAKVKSLLNKDLSLENGAHAYN | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
8E3H1 | |
ChIP Assay, Immunofluorescence, Immunoprecipitation, KnockDown | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Stem Cell Markers, Transcription Factors and Regulators | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1786 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title