Learn More
domain-II-of-FIZ1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15827-1-AP-150UL
Description
FLJ14768 is homologous to mouse and bovine Fiz1 (flt-3 interacting zinc finger). It interacts with NRL (neural retina leucine zipper). Possible role for Fiz1 as a transcriptional repressor.Specifications
domain-II-of-FIZ1 | |
Polyclonal | |
Unconjugated | |
FIZ1 | |
domain I of FIZ 1, domain II of FIZ1, FIZ1, FLT3 interacting zinc finger 1, Zinc finger protein 798, ZNF798 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84922 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96SL8 | |
FIZ1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.