Learn More
Sampling Systems™ Double Bagged - USP Class VI PharmaScoops™
BIOPHARMA
Double Bagged PharmaScoops™ are made from high grade medical USP Class VI HDPE. They are available in a range of sizes from 125 mL to 1000 mL. This premium range is double bagged. A box liner is also used giving a third layer of protection.
1400.00 SEK - 2090.00 SEK
Specifications
Material HDPE
Sterility Sterile
Packaging Type Double Bagged
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17544696
BIOPHARMA
View Documents
Sampling Systems™
9045H-125S
125 mL
2090.00 SEK
Pack of 50
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
17554696
BIOPHARMA
View Documents
Sampling Systems™
9045H-250S
250 mL
1590.00 SEK
Pack of 25
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
17534696
BIOPHARMA
View Documents
Sampling Systems™
9045H-1000S
1000 mL
1400.00 SEK
Pack of 10
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
  • These sterile scoops are commonly used in the manufacture and sampling of cosmetics, food and pharmaceuticals.
  • These scoops all have an ergonomic handle which is positioned high up the body so as to keep the hand as far away from the powder as possible.
  • The scoops are designed to be used straight from the bag.
  • The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuring the cleanliness inside the cleanroom or manufacturing area.
  • Made from food grade, FDA and EC compliant HDPE.
  • HDPE has good chemical resistance and can be easily recycled.
  • The funnels are sterilised by gamma irradiation.
  • USP Class VI - High quality medical grade polymer that has also been independently tested to USP 661.
  • The scoops are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
  • Each scoop is 100% inspected before packing.
Specifications
Specifications
HDPE
Sterile
Double Bagged